Product Details

SNP ID
rs11065215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120692543 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCGTGGCTAATGTCACTTACAGGA[A/C]CTGTTAACAGTTATGTTTATTTTTT
Phenotype
MIM: 613802
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MLEC PubMed Links

Gene Details

Gene
MLEC
Gene Name
malectin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303627.1 4238 Intron NP_001290556.1
NM_001303628.1 4238 Intron NP_001290557.1
NM_014730.3 4238 Intron NP_055545.1
XM_011539031.1 4238 UTR 5 XP_011537333.1
XM_011539032.1 4238 Intron XP_011537334.1

View Full Product Details