Product Details
- SNP ID
-
rs12170238
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:20445128 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACTACAATCAGGCAGAAAACATAC[A/C]CGGTCAATGCTGAAAATGGTCATAC
- Phenotype
-
MIM: 613619
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KLHL22
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114552752] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KLHL22
- Gene Name
- kelch like family member 22
- Gene
- SCARF2
- Gene Name
- scavenger receptor class F member 2
There are no transcripts associated with this gene.
View Full Product Details