Product Details

SNP ID
rs11570432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38521257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATTTAACACAATTTAATTGGAAT[C/T]GATGATTTCATTATCTAGGTGTATG
Phenotype
MIM: 602721
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DMC1 PubMed Links

Gene Details

Gene
DMC1
Gene Name
DNA meiotic recombinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278208.1 Intron NP_001265137.1
NM_007068.3 Intron NP_008999.2
XM_006724112.2 Intron XP_006724175.1
XM_011529834.1 Intron XP_011528136.1
XM_011529835.2 Intron XP_011528137.1
XM_011529837.1 Intron XP_011528139.1
XM_011529838.1 Intron XP_011528140.1
XM_017028542.1 Intron XP_016884031.1

View Full Product Details