Product Details
- SNP ID
-
rs12097502
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
7
- Location
-
Chr.1:234759807 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCCCTGCCTTCAAGAAGAGCAGTT[C/G]CTCTTCCATCTGCTTGAGGCATTGG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PP2672
PubMed Links
Gene Details
- Gene
- PP2672
- Gene Name
- uncharacterized LOC100130249
There are no transcripts associated with this gene.
View Full Product Details