Product Details

SNP ID
rs12802850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77132316 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGACATGAGGGACAGCCTTCATCC[C/T]ATTTCCCGTGGATCTTTCAGGATAG
Phenotype
MIM: 602537 MIM: 276903
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAPN5 PubMed Links

Gene Details

Gene
CAPN5
Gene Name
calpain 5
There are no transcripts associated with this gene.

Gene
MYO7A
Gene Name
myosin VIIA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000260.3 Intron NP_000251.3
NM_001127179.2 Intron NP_001120651.2
NM_001127180.1 Intron NP_001120652.1
XM_011545044.2 Intron XP_011543346.1
XM_011545046.2 Intron XP_011543348.2
XM_011545050.2 Intron XP_011543352.1
XM_017017778.1 Intron XP_016873267.1
XM_017017779.1 Intron XP_016873268.1
XM_017017780.1 Intron XP_016873269.1
XM_017017781.1 Intron XP_016873270.1
XM_017017782.1 Intron XP_016873271.1
XM_017017783.1 Intron XP_016873272.1
XM_017017784.1 Intron XP_016873273.1
XM_017017785.1 Intron XP_016873274.1
XM_017017786.1 Intron XP_016873275.1
XM_017017787.1 Intron XP_016873276.1
XM_017017788.1 Intron XP_016873277.1

View Full Product Details