Product Details

SNP ID
rs11705868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129975270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTTAGCATTAGAAATAGAATCT[C/T]AGCTTTGCAAATAGACTTTGAGCTT
Phenotype
MIM: 613879
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRH PubMed Links

Gene Details

Gene
TRH
Gene Name
thyrotropin releasing hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007117.4 Intron NP_009048.1

View Full Product Details