Product Details

SNP ID
rs11693889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:28400140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGAGGAGGACATGTTGGCTACA[A/G]AGCCCCAGGCAGCCGAGTGAGGTTT
Phenotype
MIM: 601575
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLJ31356 PubMed Links

Gene Details

Gene
FLJ31356
Gene Name
uncharacterized protein FLJ31356
There are no transcripts associated with this gene.

Gene
FOSL2
Gene Name
FOS like 2, AP-1 transcription factor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005253.3 Intron NP_005244.1
XM_005264231.3 Intron XP_005264288.1
XM_006711976.2 Intron XP_006712039.1
XM_006711977.3 Intron XP_006712040.1
XM_017003737.1 Intron XP_016859226.1

View Full Product Details