Product Details

SNP ID
rs13033383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:97723050 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCCTGTGAATATACAGGACGTCA[C/T]CATCACCCCAGAAAGTTGAGGGCAG
Phenotype
MIM: 176947
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZAP70 PubMed Links

Gene Details

Gene
ZAP70
Gene Name
zeta chain of T cell receptor associated protein kinase 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079.3 Intron NP_001070.2
NM_207519.1 Intron NP_997402.1
XM_017004867.1 Intron XP_016860356.1
XM_017004868.1 Intron XP_016860357.1
XM_017004869.1 Intron XP_016860358.1
XM_017004870.1 Intron XP_016860359.1

View Full Product Details