Product Details

SNP ID
rs7031160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:71687880 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTTCTTTTGTTTTTTTAGAGATA[A/G]GGTCCTTGCTCTGTCACCCCAGCTG
Phenotype
MIM: 605835
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM2 PubMed Links

Gene Details

Gene
TMEM2
Gene Name
transmembrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135820.1 Intron NP_001129292.1
NM_013390.2 Intron NP_037522.1
XM_005251869.4 Intron XP_005251926.1

View Full Product Details