Product Details

SNP ID
rs12224729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126214046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGTGATTTAAAGTAGAGTTGTT[C/T]TCAGTATAAAGAATTTATGTGGCCG
Phenotype
MIM: 616587
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM118B PubMed Links

Gene Details

Gene
FAM118B
Gene Name
family with sequence similarity 118 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024556.3 Intron NP_078832.1
XM_005271665.3 Intron XP_005271722.1
XM_011542977.2 Intron XP_011541279.1
XM_011542978.2 Intron XP_011541280.1
XM_017018285.1 Intron XP_016873774.1
XM_017018286.1 Intron XP_016873775.1
Gene
RPUSD4
Gene Name
RNA pseudouridylate synthase domain containing 4
There are no transcripts associated with this gene.

View Full Product Details