Product Details

SNP ID
rs11562977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233917411 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCCTTGGGTGAAAGAAAATCCTGC[C/T]TGACAAAAACCGTCACTTAGGGTAT
Phenotype
MIM: 606678
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRPM8 PubMed Links

Gene Details

Gene
TRPM8
Gene Name
transient receptor potential cation channel subfamily M member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024080.4 14 UTR 5 NP_076985.4
XM_011511810.2 14 UTR 5 XP_011510112.1
XM_017004891.1 14 Intron XP_016860380.1

View Full Product Details