Product Details

SNP ID
rs6600158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1622601 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGCGATTTGCTTTCTTGTCTGTT[G/T]TTTGGGGTTTTCTGTCTGTTTTGTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CRAMP1 PubMed Links

Gene Details

Gene
CRAMP1
Gene Name
cramped chromatin regulator homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020825.3 Intron NP_065876.3

View Full Product Details