Product Details

SNP ID
rs11111135
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:102023379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTATATGAGTTTGTCTTACTAC[C/G]TAAAGCTAATAAAAAATAAAGTGTA
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CCDC53 PubMed Links

Gene Details

Gene
CCDC53
Gene Name
coiled-coil domain containing 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301107.1 Intron NP_001288036.1
NM_016053.3 Intron NP_057137.1
XM_011538423.1 Intron XP_011536725.1
XM_017019383.1 Intron XP_016874872.1

View Full Product Details