Product Details
- SNP ID
-
rs4150060
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:77159810 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATAGTCCTCAATATAAGATTTCAT[C/T]TGCCATCATGCTTCTCTTGAAGCCA
- Phenotype
-
MIM: 603203
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCNG2
PubMed Links
Gene Details
- Gene
- CCNG2
- Gene Name
- cyclin G2
View Full Product Details