Product Details

SNP ID
rs4660848
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:45331176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGTACAACAAAGACAACAAAGG[G/T]AGTGCCTTTTTCATGGCGGTGGAAA
Phenotype
MIM: 604933 MIM: 613931
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HPDL PubMed Links

Gene Details

Gene
HPDL
Gene Name
4-hydroxyphenylpyruvate dioxygenase like
There are no transcripts associated with this gene.

Gene
MUTYH
Gene Name
mutY DNA glycosylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048171.1 Intron NP_001041636.1
NM_001048172.1 Intron NP_001041637.1
NM_001048173.1 Intron NP_001041638.1
NM_001048174.1 Intron NP_001041639.1
NM_001128425.1 Intron NP_001121897.1
NM_001293190.1 Intron NP_001280119.1
NM_001293191.1 Intron NP_001280120.1
NM_001293192.1 Intron NP_001280121.1
NM_001293195.1 Intron NP_001280124.1
NM_001293196.1 Intron NP_001280125.1
NM_012222.2 Intron NP_036354.1
XM_011541497.2 Intron XP_011539799.1
XM_011541498.1 Intron XP_011539800.1
XM_011541499.1 Intron XP_011539801.1
XM_011541500.2 Intron XP_011539802.1
XM_011541501.2 Intron XP_011539803.1
XM_011541502.2 Intron XP_011539804.1
XM_011541503.2 Intron XP_011539805.1
XM_011541504.2 Intron XP_011539806.1
XM_011541505.2 Intron XP_011539807.1
XM_011541506.1 Intron XP_011539808.1
XM_011541507.2 Intron XP_011539809.2
XM_017001331.1 Intron XP_016856820.1
XM_017001332.1 Intron XP_016856821.1
XM_017001333.1 Intron XP_016856822.1
XM_017001334.1 Intron XP_016856823.1
XM_017001335.1 Intron XP_016856824.1
XM_017001336.1 Intron XP_016856825.1
XM_017001337.1 Intron XP_016856826.1
XM_017001338.1 Intron XP_016856827.1
XM_017001339.1 Intron XP_016856828.1
Gene
TOE1
Gene Name
target of EGR1, member 1 (nuclear)
There are no transcripts associated with this gene.

View Full Product Details