Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_012296.3 | 5973 | UTR 3 | NP_036428.1 | ||
NM_080491.2 | 5973 | UTR 3 | NP_536739.1 | ||
XM_006718753.2 | 5973 | UTR 3 | XP_006718816.1 | ||
XM_011545408.2 | 5973 | UTR 3 | XP_011543710.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_020798.2 | 5973 | Intron | NP_065849.1 | ||
XM_011545187.2 | 5973 | Intron | XP_011543489.1 | ||
XM_011545188.2 | 5973 | Intron | XP_011543490.1 | ||
XM_017018052.1 | 5973 | Intron | XP_016873541.1 | ||
XM_017018053.1 | 5973 | Intron | XP_016873542.1 |