Product Details

SNP ID
rs17869893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:90212924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTAAACCACCTCCCCATCCTTC[A/C]TTTTCTCCACTTTATTATTTCTTCA
Phenotype
MIM: 605094
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
STEAP2 PubMed Links

Gene Details

Gene
STEAP2
Gene Name
STEAP2 metalloreductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040665.1 456 Intron NP_001035755.1
NM_001040666.1 456 Intron NP_001035756.1
NM_001244944.1 456 Intron NP_001231873.1
NM_001244945.1 456 Intron NP_001231874.1
NM_001244946.1 456 Intron NP_001231875.1
NM_152999.3 456 Intron NP_694544.2
XM_006715921.3 456 Intron XP_006715984.1
XM_017011953.1 456 Intron XP_016867442.1
XM_017011954.1 456 Intron XP_016867443.1
XM_017011955.1 456 Intron XP_016867444.1
XM_017011956.1 456 Intron XP_016867445.1
XM_017011957.1 456 Intron XP_016867446.1
XM_017011958.1 456 UTR 5 XP_016867447.1
Gene
STEAP2-AS1
Gene Name
STEAP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details