Product Details

SNP ID
rs17867591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:90216235 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGAATCTGAGTGTGCATTTTAAA[A/G]AGGTCCCCAGGTGATCATATTTGCA
Phenotype
MIM: 605094
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STEAP2 PubMed Links

Gene Details

Gene
STEAP2
Gene Name
STEAP2 metalloreductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040665.1 3767 Intron NP_001035755.1
NM_001040666.1 3767 Intron NP_001035756.1
NM_001244944.1 3767 Intron NP_001231873.1
NM_001244945.1 3767 Intron NP_001231874.1
NM_001244946.1 3767 Intron NP_001231875.1
NM_152999.3 3767 Intron NP_694544.2
XM_006715921.3 3767 Intron XP_006715984.1
XM_017011953.1 3767 Intron XP_016867442.1
XM_017011954.1 3767 Intron XP_016867443.1
XM_017011955.1 3767 Intron XP_016867444.1
XM_017011956.1 3767 Intron XP_016867445.1
XM_017011957.1 3767 Intron XP_016867446.1
XM_017011958.1 3767 UTR 5 XP_016867447.1
Gene
STEAP2-AS1
Gene Name
STEAP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details