Product Details

SNP ID
rs16858198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:142883541 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTTCTGCCTTTGTTTTTTGCATA[G/T]CACAGTTTGTCATTCCTCCTTCCCT
Phenotype
MIM: 605197
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KYNU PubMed Links

Gene Details

Gene
KYNU
Gene Name
kynureninase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032998.1 Intron NP_001028170.1
NM_001199241.1 Intron NP_001186170.1
NM_003937.2 Intron NP_003928.1
XM_011512102.2 Intron XP_011510404.1
XM_017005217.1 Intron XP_016860706.1

View Full Product Details