Product Details

SNP ID
rs16868542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:11544438 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAGTACCCTGGTAAGTGTTTGCA[G/T]CAGTGAGCCTGAAGATGTATGAATG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM170B PubMed Links

Gene Details

Gene
TMEM170B
Gene Name
transmembrane protein 170B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100829.2 Intron NP_001094299.1

View Full Product Details