Product Details

SNP ID
rs35818504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32452546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTAGAAGACAGAATTAAAATTC[A/C]CAGTTATTGCCAAAGCCAGGAAGAT
Phenotype
MIM: 604449
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PSMD11 PubMed Links

Gene Details

Gene
PSMD11
Gene Name
proteasome 26S subunit, non-ATPase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270482.1 Intron NP_001257411.1
NM_002815.3 Intron NP_002806.2

View Full Product Details