Product Details

SNP ID
rs17032220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:102013968 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTCATTTAATCAGTGTAGTGGCA[C/T]TATCACTTAGCCTCTATTCACTCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC53 PubMed Links

Gene Details

Gene
CCDC53
Gene Name
coiled-coil domain containing 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301107.1 Intron NP_001288036.1
NM_016053.3 Intron NP_057137.1
XM_011538423.1 Intron XP_011536725.1
XM_017019383.1 Intron XP_016874872.1

View Full Product Details