Product Details

SNP ID
rs17663627
Assay Type
Functionally Tested
NCBI dbSNP Submissions
17
Location
Chr.6:49510558 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTTCTATCAGGATATATTGAATT[C/T]GAATAAATAAATGTATTTCAAATAA
Phenotype
MIM: 614763
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GLYATL3 PubMed Links

Gene Details

Gene
GLYATL3
Gene Name
glycine-N-acyltransferase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010904.1 Intron NP_001010904.1

View Full Product Details