Product Details

SNP ID
rs8002671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:19965164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAGTTAGAAGTTGCAGACAGAA[C/T]AATGTTTGTAAAATGTGACTATAAT
Phenotype
MIM: 602221
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZMYM2 PubMed Links

Gene Details

Gene
ZMYM2
Gene Name
zinc finger MYM-type containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190964.2 Intron NP_001177893.1
NM_001190965.2 Intron NP_001177894.1
NM_003453.4 Intron NP_003444.1
NM_197968.3 Intron NP_932072.1
XM_005266520.3 Intron XP_005266577.1
XM_011535216.2 Intron XP_011533518.1
XM_011535222.1 Intron XP_011533524.1
XM_011535223.2 Intron XP_011533525.1
XM_011535224.2 Intron XP_011533526.1
XM_017020728.1 Intron XP_016876217.1
XM_017020729.1 Intron XP_016876218.1
XM_017020731.1 Intron XP_016876220.1
XM_017020732.1 Intron XP_016876221.1
XM_017020733.1 Intron XP_016876222.1
XM_017020734.1 Intron XP_016876223.1
XM_017020735.1 Intron XP_016876224.1
XM_017020736.1 Intron XP_016876225.1
XM_017020737.1 Intron XP_016876226.1
XM_017020738.1 Intron XP_016876227.1
XM_017020739.1 Intron XP_016876228.1

View Full Product Details