Product Details

SNP ID
rs11537864
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:44777667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGTCTACAATGCATCTAATAAC[G/T]AGCTGGTTCGTACCAAGACCCTGGT
Phenotype
MIM: 607336 MIM: 604538 MIM: 600357
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BEST4 PubMed Links

Gene Details

Gene
BEST4
Gene Name
bestrophin 4
There are no transcripts associated with this gene.

Gene
KIF2C
Gene Name
kinesin family member 2C
There are no transcripts associated with this gene.

Gene
RPS8
Gene Name
ribosomal protein S8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012.1 Intron NP_001003.1
Gene
SNORD38A
Gene Name
small nucleolar RNA, C/D box 38A
There are no transcripts associated with this gene.

Gene
SNORD38B
Gene Name
small nucleolar RNA, C/D box 38B
There are no transcripts associated with this gene.

Gene
SNORD46
Gene Name
small nucleolar RNA, C/D box 46
There are no transcripts associated with this gene.

Gene
SNORD55
Gene Name
small nucleolar RNA, C/D box 55
There are no transcripts associated with this gene.

View Full Product Details