Product Details

SNP ID
rs7477921
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:21534596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGGGCTGTGTGGGGGGGAAGCA[A/C]TAATCGGCTTGCATGTGTTTTTTAA
Phenotype
MIM: 602409
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
MLLT10 PubMed Links

Gene Details

Gene
MLLT10
Gene Name
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195626.1 Intron NP_001182555.1
NM_001195627.1 Intron NP_001182556.1
NM_001195628.1 Intron NP_001182557.1
NM_001195630.1 Intron NP_001182559.1
NM_001324296.1 Intron NP_001311225.1
NM_001324297.1 Intron NP_001311226.1
NM_004641.3 Intron NP_004632.1
Gene
SKIDA1
Gene Name
SKI/DACH domain containing 1
There are no transcripts associated with this gene.

View Full Product Details