Product Details

SNP ID
rs11553049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:44715537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGACATTGAAGTTGACTTACTGA[A/G]GAATGGAGAGAGAATTGAAAAAGTG
Phenotype
MIM: 109700 MIM: 614661
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B2M PubMed Links

Gene Details

Gene
B2M
Gene Name
beta-2-microglobulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004048.2 237 Missense Mutation AAG,AGG K61R NP_004039.1
XM_005254549.3 237 Missense Mutation AAG,AGG K61R XP_005254606.1
Gene
PATL2
Gene Name
PAT1 homolog 2
There are no transcripts associated with this gene.

View Full Product Details