Product Details

SNP ID
rs6653263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:68655306 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGACAGTATATACAGCCAGCCAC[A/G]TCTTAATTACTTGTTCCAAAGAAAT
Phenotype
MIM: 300689
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STARD8 PubMed Links

Gene Details

Gene
STARD8
Gene Name
StAR related lipid transfer domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142503.2 Intron NP_001135975.1
NM_001142504.2 Intron NP_001135976.1
NM_014725.4 Intron NP_055540.2
XM_005262314.4 Intron XP_005262371.1
XM_005262315.3 Intron XP_005262372.1
XM_011531069.2 Intron XP_011529371.1
XM_011531070.2 Intron XP_011529372.1

View Full Product Details