Product Details

SNP ID
rs11538959
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:112702613 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGTGTGCTCGTCTTGCCTCAGGT[C/G]CTTCTTATTCCCCACCAGGATGATG
Phenotype
MIM: 610742 MIM: 609957 MIM: 165380
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MOV10 PubMed Links

Gene Details

Gene
MOV10
Gene Name
Mov10 RISC complex RNA helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130079.2 691 Intron NP_001123551.1
NM_001286072.1 691 Intron NP_001273001.1
NM_001321324.1 691 Intron NP_001308253.1
NM_020963.4 691 Intron NP_066014.1
XM_005270869.4 691 Intron XP_005270926.1
XM_005270870.4 691 Intron XP_005270927.1
XM_017001318.1 691 Intron XP_016856807.1
XM_017001319.1 691 Intron XP_016856808.1
Gene
PPM1J
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1J
There are no transcripts associated with this gene.

Gene
RHOC
Gene Name
ras homolog family member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042678.1 691 Missense Mutation CAC,GAC H120D NP_001036143.1
NM_001042679.1 691 Missense Mutation CAC,GAC H120D NP_001036144.1
NM_175744.4 691 Missense Mutation CAC,GAC H120D NP_786886.1

View Full Product Details