Product Details

SNP ID
rs2688476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:195731852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGCCAATCGTGACTCCCACTGT[A/G]AGGAGTTCCCTAAACATGGCACGGG
Phenotype
MIM: 610360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MUC20 PubMed Links

Gene Details

Gene
MUC20
Gene Name
mucin 20, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282506.1 Intron NP_001269435.1
NM_001291833.1 Intron NP_001278762.1
NM_020790.1 Intron NP_065841.1
NM_152673.3 Intron NP_689886.3

View Full Product Details