Product Details

SNP ID
rs2813454
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:1529935 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCCACTGAACCCTGCCACTGCAGG[C/G]ACCCATTCACTGCCCCTGCCACTGT
Phenotype
MIM: 602065
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ADARB2 PubMed Links

Gene Details

Gene
ADARB2
Gene Name
adenosine deaminase, RNA specific B2 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018702.3 Intron NP_061172.1
Gene
ADARB2-AS1
Gene Name
ADARB2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details