Product Details

SNP ID
rs4648793
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:1740343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCTACTTTCTTTTTTTTTGAGA[C/T]GGGAGTCTCGCTGTGTCGCCTAGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC35E2 PubMed Links

Gene Details

Gene
SLC35E2
Gene Name
solute carrier family 35 member E2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199787.1 Intron NP_001186716.1
NM_182838.2 Intron NP_878258.1

View Full Product Details