Product Details

SNP ID
rs1804743
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:154962325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAAATACGGACTTTAATGAAGA[A/G]TTTTCCCTTTGGTATAAGTGAGACC
Phenotype
MIM: 606903 MIM: 600560
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928120 PubMed Links

Gene Details

Gene
LOC101928120
Gene Name
uncharacterized LOC101928120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321726.1 855 UTR 3 NP_001308655.1
Gene
PBXIP1
Gene Name
PBX homeobox interacting protein 1
There are no transcripts associated with this gene.

Gene
PYGO2
Gene Name
pygopus family PHD finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138300.3 855 Intron NP_612157.1
Gene
SHC1
Gene Name
SHC adaptor protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130040.1 855 Intron NP_001123512.1
NM_001130041.1 855 Intron NP_001123513.1
NM_001202859.1 855 Intron NP_001189788.1
NM_003029.4 855 Intron NP_003020.2
NM_183001.4 855 Intron NP_892113.4
XM_005245449.4 855 Intron XP_005245506.1
XM_005245451.4 855 Intron XP_005245508.1
XM_011509892.2 855 UTR 3 XP_011508194.1
XM_011509893.2 855 UTR 3 XP_011508195.1
XM_011509894.2 855 UTR 3 XP_011508196.1
XM_011509897.1 855 Intron XP_011508199.1
XM_011509898.2 855 Intron XP_011508200.1
XM_017002081.1 855 UTR 3 XP_016857570.1
XM_017002082.1 855 UTR 3 XP_016857571.1
XM_017002083.1 855 Intron XP_016857572.1

View Full Product Details