Product Details

SNP ID
rs3210097
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:26191100 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTTACAGTCTTTGTGGCAGCAG[A/T]ATATACTTGTCCATGGTTCATATCA
Phenotype
MIM: 600124 MIM: 604135
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
HNRNPA2B1 PubMed Links

Gene Details

Gene
HNRNPA2B1
Gene Name
heterogeneous nuclear ribonucleoprotein A2/B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002137.3 2538 UTR 3 NP_002128.1
NM_031243.2 2538 UTR 3 NP_112533.1
XM_005249729.1 2538 Intron XP_005249786.1
XM_017012109.1 2538 UTR 3 XP_016867598.1
XM_017012110.1 2538 Intron XP_016867599.1
Gene
NFE2L3
Gene Name
nuclear factor, erythroid 2 like 3
There are no transcripts associated with this gene.

View Full Product Details