Product Details

SNP ID
rs28582122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:33728466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTTCTGAATTAAATAGTTCCTG[C/T]CTGGTAATTTGTATCAACTGAGGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLJ20712 PubMed Links

Gene Details

Gene
FLJ20712
Gene Name
uncharacterized FLJ20712
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017012879.1 2020 UTR 3 XP_016868368.1

View Full Product Details