Product Details

SNP ID
rs35470760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:117485152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATAAAATGCATTATTATTGCTAG[G/T]GTCATTTCACAAGAGCCTATAATTT
Phenotype
MIM: 602421
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CFTR PubMed Links

Gene Details

Gene
CFTR
Gene Name
cystic fibrosis transmembrane conductance regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000492.3 Intron NP_000483.3
XM_011515751.2 Intron XP_011514053.1
XM_011515753.1 Intron XP_011514055.1
XM_011515754.2 Intron XP_011514056.1
XM_017011699.1 Intron XP_016867188.1

View Full Product Details