Product Details

SNP ID
rs28759168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75177080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTGTAGTCCCAGCTACTCCAGC[C/T]GGAGGTTGAGGGGGGAAAATTGCAT
Phenotype
MIM: 603042
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUMO2 PubMed Links

Gene Details

Gene
SUMO2
Gene Name
small ubiquitin-like modifier 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005849.1 Intron NP_001005849.1
NM_006937.3 Intron NP_008868.3

View Full Product Details