Product Details

SNP ID
rs28478232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11203870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGAGAGCTCCCAGCTGGGTGA[A/G]TCACAAGCTGCCCTCGAGATCTGGG
Phenotype
MIM: 614194 MIM: 614610
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DOCK6 PubMed Links

Gene Details

Gene
DOCK6
Gene Name
dedicator of cytokinesis 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020812.3 Intron NP_065863.2
XM_005260000.2 Intron XP_005260057.1
XM_005260001.2 Intron XP_005260058.1
XM_006722804.3 Intron XP_006722867.1
XM_011528150.1 Intron XP_011526452.1
XM_011528151.1 Intron XP_011526453.1
XM_011528152.1 Intron XP_011526454.1
Gene
KANK2
Gene Name
KN motif and ankyrin repeat domains 2
There are no transcripts associated with this gene.

Gene
LOC105372273
Gene Name
uncharacterized LOC105372273
There are no transcripts associated with this gene.

View Full Product Details