Product Details

SNP ID
rs28647355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24938800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTTTGGCAATGTGCATCATAGC[G/T]GGGACTTTAACACCTTAGCTTTCAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM211 PubMed Links

Gene Details

Gene
TMEM211
Gene Name
transmembrane protein 211
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001663.1 378 Intron NP_001001663.1
XM_011530094.2 378 UTR 5 XP_011528396.1

View Full Product Details