Product Details

SNP ID
rs879075989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:87894269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAATTAAAGAATGTAAAATAAA[A/G]AATATGCCGTTTTTATACAAAAAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSP90AB3P PubMed Links

Gene Details

Gene
HSP90AB3P
Gene Name
heat shock protein 90 alpha family class B member 3, pseudogene
There are no transcripts associated with this gene.

View Full Product Details