Product Details

SNP ID
rs11541609
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.1:10946733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGGGCTGTCCAAGCCAGCGGGG[C/T]ACCGGCTGCTGCCATGGAACCCAGA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C1orf127 PubMed Links

Gene Details

Gene
C1orf127
Gene Name
chromosome 1 open reading frame 127
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170754.1 2419 UTR 3 NP_001164225.1
XM_017000373.1 2419 UTR 3 XP_016855862.1

View Full Product Details