Product Details

SNP ID
rs28411135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77322797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTTGGAAGGGCCTAGGTTCCCA[C/T]TTACACCCTTGCCATCTAGCAGACT
Phenotype
MIM: 603758 MIM: 607439
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTZ1 PubMed Links

Gene Details

Gene
GSTZ1
Gene Name
glutathione S-transferase zeta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001312660.1 Intron NP_001299589.1
NM_145870.2 Intron NP_665877.1
NM_145871.2 Intron NP_665878.2
XM_011536670.2 Intron XP_011534972.1
XM_011536671.2 Intron XP_011534973.1
XM_017021243.1 Intron XP_016876732.1
Gene
POMT2
Gene Name
protein O-mannosyltransferase 2
There are no transcripts associated with this gene.

View Full Product Details