Product Details

SNP ID
rs34538771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:41688232 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGAGTGTACCCAGAAATAATTAG[A/G]ATGTTAGATACAGCCCAATGAGTCA
Phenotype
MIM: 300172 MIM: 300241
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASK PubMed Links

Gene Details

Gene
CASK
Gene Name
calcium/calmodulin dependent serine protein kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126054.2 1690 Intron NP_001119526.1
NM_001126055.2 1690 Intron NP_001119527.1
NM_003688.3 1690 Intron NP_003679.2
XM_005272686.4 1690 Intron XP_005272743.1
XM_006724566.3 1690 Intron XP_006724629.1
XM_011543993.2 1690 Intron XP_011542295.1
XM_011543994.2 1690 Intron XP_011542296.1
XM_011543995.2 1690 Intron XP_011542297.1
XM_011543996.2 1690 Intron XP_011542298.1
XM_011543997.2 1690 Intron XP_011542299.1
Gene
GPR34
Gene Name
G protein-coupled receptor 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097579.1 1690 Intron NP_001091048.1
NM_005300.3 1690 Intron NP_005291.1
XM_005272597.3 1690 UTR 5 XP_005272654.1

View Full Product Details