Product Details

SNP ID
rs151254333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:50668438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGTTAGGGTGTAATAACAGTGAT[C/G]GGTTTCATTGTGTTTATTTTTGTAG
Phenotype
MIM: 613281
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927272 PubMed Links

Gene Details

Gene
LOC101927272
Gene Name
uncharacterized LOC101927272
There are no transcripts associated with this gene.

Gene
SNX20
Gene Name
sorting nexin 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144972.1 1166 Intron NP_001138444.1
NM_153337.2 1166 UTR 3 NP_699168.1
NM_182854.3 1166 Intron NP_878274.1

View Full Product Details