Product Details

SNP ID
rs114052958
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.2:233682354 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
GAAAATTAGTAGAATACTTAAAGGA[C/G]AGTTGTTTTGATGCAGTGTTTCTCG
Phenotype
MIM: 606435 MIM: 606431 MIM: 606432 MIM: 606433 MIM: 606434
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
UGT1A7*6,g.417G>C UGT1A7*7,g.417G>C UGT1A7*8,g.417G>C
Literature Links
UGT1A10 PubMed Links

Gene Details

Gene
UGT1A10
Gene Name
UDP glucuronosyltransferase family 1 member A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019075.2 417 Intron NP_061948.1
Gene
UGT1A6
Gene Name
UDP glucuronosyltransferase family 1 member A6
There are no transcripts associated with this gene.

Gene
UGT1A7
Gene Name
UDP glucuronosyltransferase family 1 member A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019077.2 417 Missense Mutation GAC,GAG D139E NP_061950.2
Gene
UGT1A8
Gene Name
UDP glucuronosyltransferase family 1 member A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019076.4 417 Intron NP_061949.3
Gene
UGT1A9
Gene Name
UDP glucuronosyltransferase family 1 member A9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021027.2 417 Intron NP_066307.1

View Full Product Details