Product Details

SNP ID
rs45450891
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:72755490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAACAACAAATTGTGACAGAGCTG[A/T]GTGCTCCTATCTTACAGGGTCAATG
Phenotype
MIM: 610510 MIM: 607533
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
BORA PubMed Links

Gene Details

Gene
BORA
Gene Name
bora, aurora kinase A activator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286746.1 2228 UTR 3 NP_001273675.1
NM_001286747.1 2228 UTR 3 NP_001273676.1
NM_024808.3 2228 UTR 3 NP_079084.3
XM_006719868.3 2228 Intron XP_006719931.2
Gene
DIS3
Gene Name
DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128226.2 2228 UTR 3 NP_001121698.1
NM_001322348.1 2228 UTR 3 NP_001309277.1
NM_001322349.1 2228 UTR 3 NP_001309278.1
NM_014953.4 2228 UTR 3 NP_055768.3

View Full Product Details