Product Details

SNP ID
rs45565136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73974707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCTTTATGGATGACAAGTGGGC[G/T]AGGCTGGGCTGTCGGGGGCAGGACG
Phenotype
MIM: 610263 MIM: 601693
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJB13 PubMed Links

Gene Details

Gene
DNAJB13
Gene Name
DnaJ heat shock protein family (Hsp40) member B13
There are no transcripts associated with this gene.

Gene
UCP2
Gene Name
uncoupling protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003355.2 1610 UTR 3 NP_003346.2

View Full Product Details