Product Details

SNP ID
rs60165033
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:160372529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTCAACTCCTGGTGTGTGTGA[A/G]CATGCCCTTTGCTTGCCACACCATA
Phenotype
MIM: 162360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NHLH1 PubMed Links

Gene Details

Gene
NHLH1
Gene Name
nescient helix-loop-helix 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005598.3 2240 UTR 3 NP_005589.1

View Full Product Details