Product Details

SNP ID
rs56270745
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:241532486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAAAGGAAAAAAGTAGCTGTCAT[A/T]GGTGGTGGCTTGGTAAGAATTTTCA
Phenotype
MIM: 603538
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KMO PubMed Links

Gene Details

Gene
KMO
Gene Name
kynurenine 3-monooxygenase (kynurenine 3-hydroxylase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003679.4 463 Silent Mutation ATA,ATT I14I NP_003670.2
XM_005273337.1 463 Silent Mutation ATA,ATT I54I XP_005273394.1
XM_005273338.3 463 UTR 5 XP_005273395.1
XM_017002649.1 463 UTR 5 XP_016858138.1
XM_017002650.1 463 Intron XP_016858139.1

View Full Product Details