Product Details
- SNP ID
-
hCV89196242
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:44711600 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGCGCTACTCTCTCTTTCTGGCCT[C/G]GAGGCTATCCAGCGTGAGTCTCTCC
- Phenotype
-
MIM: 109700
MIM: 614661
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
B2M
PubMed Links
Gene Details
- Gene
- B2M
- Gene Name
- beta-2-microglobulin
There are no transcripts associated with this gene.
- Gene
- PATL2
- Gene Name
- PAT1 homolog 2
There are no transcripts associated with this gene.
View Full Product Details